Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem55a
Entrez Gene ID
362490
Gene Name
transmembrane protein 55A
Gene Aliases
RGD1306225
Chromosome Location
Chr.5: 28395517 - 28442924 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024900.1 NP_001020071.1 6 - 7 694 91
GenBank mRNA BC097492.1 - 6 - 7 694 91

View Full Product Details