Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem8b
Entrez Gene ID
313490
Gene Name
transmembrane protein 8B
Gene Aliases
RGD1310012
Chromosome Location
Chr.5: 59165712 - 59193772 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006238139.2 XP_006238201.1 12 - 13 2507 125
XM_008763686.1 XP_008761908.1 10 - 11 3645 125
XM_008763688.1 XP_008761910.1 13 - 14 2033 125

View Full Product Details