Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tyw5
Entrez Gene ID
301419
Gene Name
tRNA-yW synthesizing protein 5
Gene Aliases
C2orf60, RGD1311269
Chromosome Location
Chr.9: 64077841 - 64096265 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170473.1 NP_001163944.1 7 - 8 962 128
NR_033170.1 7 - 8 943 128
XM_006244910.2 XP_006244972.1 6 - 7 568 128
GenBank mRNA BC158619.1 - 7 - 8 943 128

View Full Product Details