The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198789.2 | NP_942084.1 | 9 - 10 | 1204 | 68 |
XM_006245000.2 | XP_006245062.2 | 9 - 10 | 1203 | 68 | |
GenBank mRNA | BC061580.1 | - | 9 - 10 | 1208 | 68 |