Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmd5
Entrez Gene ID
296651
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 5
Gene Aliases
-
Chromosome Location
Chr.3: 13916199 - 13937038 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106569.2 NP_001100039.2 9 - 10 1284 103
XM_006233969.2 XP_006234031.1 9 - 10 1813 103
XM_006233970.2 XP_006234032.1 10 - 11 1903 103
GenBank mRNA BC166850.1 - 9 - 10 1266 103

View Full Product Details