Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcnb
Entrez Gene ID
114091
Gene Name
ficolin B
Gene Aliases
Fcn2
Chromosome Location
Chr.3: 6617816 - 6626193 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053634.1 NP_446086.1 4 - 5 311 93
GenBank mRNA AB036792.1 - 4 - 5 311 93
BC088123.1 - 4 - 5 303 93
FQ220557.1 - 4 - 5 314 93
FQ233041.1 - 4 - 5 314 93

View Full Product Details