Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ndor1
Entrez Gene ID
311799
Gene Name
NADPH dependent diflavin oxidoreductase 1
Gene Aliases
-
Chromosome Location
Chr.3: 2466702 - 2474928 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107818.1 NP_001101288.1 4 - 5 486 137
XM_006233612.2 XP_006233674.1 3 - 4 778 137

View Full Product Details