Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Edf1
Entrez Gene ID
296570
Gene Name
endothelial differentiation-related factor 1
Gene Aliases
CAP-19
Chromosome Location
Chr.3: 2781169 - 2785474 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106557.1 NP_001100027.1 1 - 2 148 88
GenBank mRNA BC158849.1 - 1 - 2 103 88

View Full Product Details