Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dctn3
Entrez Gene ID
362504
Gene Name
dynactin 3
Gene Aliases
-
Chromosome Location
Chr.5: 58098706 - 58106706 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108659.1 NP_001102129.1 3 - 4 293 59
GenBank mRNA FQ222216.1 - 3 - 4 275 59

View Full Product Details