The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106265.2 | NP_001099735.1 | 2 - 3 | 461 | 69 |
GenBank mRNA | BC166608.1 | - | 2 - 3 | 461 | 69 |