Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ifngr1
Entrez Gene ID
116465
Gene Name
interferon gamma receptor 1
Gene Aliases
Ifngr
Chromosome Location
Chr.1: 15062380 - 15080815 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC062039.1 - 2 - 3 217 139

View Full Product Details