The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053783.1 | NP_446235.1 | 4 - 5 | 611 | 99 |
GenBank mRNA | AF201901.1 | - | 4 - 5 | 611 | 99 |
BC062039.1 | - | 4 - 5 | 553 | 99 | |
U68272.1 | - | 2 - 3 | 185 | 99 |