The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276762.1 | NP_001263691.1 | 3 - 4 | 722 | 56 |
GenBank mRNA | AF347030.1 | - | - | 710 | 56 |