Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sesn1
Entrez Gene ID
294518
Gene Name
sestrin 1
Gene Aliases
-
Chromosome Location
Chr.20: 46666983 - 46758306 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006256560.2 XP_006256622.1 3 - 4 1428 69
XM_006256561.2 XP_006256623.1 3 - 4 440 69

View Full Product Details