Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bves
Entrez Gene ID
365603
Gene Name
blood vessel epicardial substance
Gene Aliases
Popdc1
Chromosome Location
Chr.20: 50439885 - 50474678 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077590.1 NP_001071058.1 3 - 4 562 83
GenBank mRNA AY786529.2 - 3 - 4 562 83

View Full Product Details