The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009700.1 | NP_001009700.1 | 8 - 9 | 1102 | 63 |
GenBank mRNA | AY383672.1 | - | - | 770 | 63 |
BC087066.1 | - | 8 - 9 | 1102 | 63 |