Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdcd2
Entrez Gene ID
58934
Gene Name
programmed cell death 2
Gene Aliases
-
Chromosome Location
Chr.1: 57512968 - 57518458 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031638.1 NP_113826.1 1 - 2 334 100
GenBank mRNA M80601.1 - 1 - 2 116 100

View Full Product Details