Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan17
Entrez Gene ID
306771
Gene Name
tetraspanin 17
Gene Aliases
Fbxo23, GHB-R
Chromosome Location
Chr.17: 10356980 - 10364605 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013138.2 NP_001013156.2 4 - 5 595 120
XM_006253630.2 XP_006253692.1 4 - 5 628 120
XM_006253631.2 XP_006253693.1 3 - 4 483 120
GenBank mRNA BC097431.1 - 4 - 5 575 120

View Full Product Details