Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asb10
Entrez Gene ID
499972
Gene Name
ankyrin repeat and SOCS box-containing 10
Gene Aliases
RGD1562922
Chromosome Location
Chr.4: 7158294 - 7167509 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109216.2 NP_001102686.1 1 - 2 398 72
GenBank mRNA BC168867.1 - 1 - 2 398 72

View Full Product Details