Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem60
Entrez Gene ID
296761
Gene Name
transmembrane protein 60
Gene Aliases
RGD1308939
Chromosome Location
Chr.4: 10818024 - 10822879 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191610.1 NP_001178539.1 1 - 2 181 126

View Full Product Details