Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tgs1
Entrez Gene ID
312947
Gene Name
trimethylguanosine synthase 1
Gene Aliases
Ncoa6ip
Chromosome Location
Chr.5: 16413356 - 16446881 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107904.1 NP_001101374.1 12 - 13 2582 143
XM_006237833.2 XP_006237895.1 12 - 13 2564 143

View Full Product Details