The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017453.1 | NP_001017453.1 | 5 - 6 | 657 | 72 |
XM_006251293.2 | XP_006251355.1 | 5 - 6 | 1866 | 72 | |
GenBank mRNA | BC092208.1 | - | 5 - 6 | 657 | 72 |