The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015022.3 | NP_001015022.1 | 8 - 9 | 1053 | 68 |
GenBank mRNA | AB210856.1 | - | 8 - 9 | 1053 | 68 |
BC093603.1 | - | 8 - 9 | 1116 | 68 |