Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bod1l1
Entrez Gene ID
207118
Gene Name
biorientation of chromosomes in cell division 1-like 1
Gene Aliases
Abp-10, Abp10, Bod1l
Chromosome Location
Chr.14: 73738137 - 73792935 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770238.1 XP_008768460.1 4 - 5 1299 107

View Full Product Details