The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013128.2 | NP_001013146.2 | 1 - 1 | 106 | 72 |
XM_006251299.2 | XP_006251361.1 | 1 - 1 | 530 | 72 | |
GenBank mRNA | BC079285.1 | - | 1 - 1 | 88 | 72 |