Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Znrf3
Entrez Gene ID
685487
Gene Name
zinc and ring finger 3
Gene Aliases
-
Chromosome Location
Chr.14: 85527099 - 85586285 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008770413.1 XP_008768635.1 3 - 4 556 73

View Full Product Details