Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pes1
Entrez Gene ID
289740
Gene Name
pescadillo ribosomal biogenesis factor 1
Gene Aliases
-
Chromosome Location
Chr.14: 84211600 - 84227980 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044228.1 NP_001037693.1 11 - 12 1367 79
GenBank mRNA BC105910.1 - 11 - 12 1367 79

View Full Product Details