Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl8
Entrez Gene ID
289457
Gene Name
kelch-like family member 8
Gene Aliases
-
Chromosome Location
Chr.14: 7169515 - 7215025 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105995.1 NP_001099465.1 4 - 5 1235 146
XM_008770004.1 XP_008768226.1 6 - 7 1570 146
XM_008770005.1 XP_008768227.1 6 - 7 1474 146
XM_008770006.1 XP_008768228.1 6 - 7 1412 146
XM_008770007.1 XP_008768229.1 5 - 6 1239 146

View Full Product Details