Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcp4
Entrez Gene ID
25510
Gene Name
Purkinje cell protein 4
Gene Aliases
PEPZ19, pep-19
Chromosome Location
Chr.11: 36851038 - 36912231 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ493658.1 - 1 - 2 61 78

View Full Product Details