Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casr
Entrez Gene ID
24247
Gene Name
calcium-sensing receptor
Gene Aliases
-
Chromosome Location
Chr.11: 67188630 - 67258721 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016996.1 NP_058692.1 4 - 5 1944 59
GenBank mRNA AY214122.1 - 4 - 5 1934 59
U10354.1 - 4 - 5 1944 59
U20289.1 - 3 - 4 1570 59

View Full Product Details