Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pax2
Entrez Gene ID
293992
Gene Name
paired box 2
Gene Aliases
-
Chromosome Location
Chr.1: 264493579 - 264583883 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106361.1 NP_001099831.1 10 - 11 1201 127
XM_008760405.1 XP_008758627.1 10 - 11 1847 127

View Full Product Details