Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pax2
Entrez Gene ID
293992
Gene Name
paired box 2
Gene Aliases
-
Chromosome Location
Chr.1: 264493579 - 264583883 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106361.1 NP_001099831.1 6 - 7 716 94
XM_006231443.2 XP_006231505.1 7 - 8 1434 94
XM_008760403.1 XP_008758625.1 7 - 8 1431 94

View Full Product Details