Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bsn
Entrez Gene ID
29138
Gene Name
bassoon (presynaptic cytomatrix protein)
Gene Aliases
Znf231
Chromosome Location
Chr.8: 116873721 - 116965396 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019146.2 NP_062019.2 9 - 10 11864 75
GenBank mRNA CB580320.1 - 3 - 4 454 75
Y16563.1 - 9 - 10 11879 75

View Full Product Details