Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mras
Entrez Gene ID
25482
Gene Name
muscle RAS oncogene homolog
Gene Aliases
-
Chromosome Location
Chr.8: 107629028 - 107656851 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012981.2 NP_037113.2 3 - 4 463 71
GenBank mRNA BC085752.1 - 3 - 4 463 71
D89863.1 - - 871 71

View Full Product Details