The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005549.1 | NP_001005549.1 | 5 - 6 | 801 | 69 |
GenBank mRNA | BC081887.1 | - | 5 - 6 | 801 | 69 |