Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exog
Entrez Gene ID
301062
Gene Name
endo/exonuclease (5'-3'), endonuclease G-like
Gene Aliases
Endogl1
Chromosome Location
Chr.8: 128133405 - 128153110 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106866.1 NP_001100336.1 2 - 3 358 80
XM_006244088.2 XP_006244150.1 3 - 4 419 80

View Full Product Details