Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nicn1
Entrez Gene ID
619581
Gene Name
nicolin 1
Gene Aliases
-
Chromosome Location
Chr.8: 117062905 - 117068136 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034999.1 NP_001030171.1 2 - 3 380 63
XM_006243854.2 XP_006243916.1 2 - 3 464 63
GenBank mRNA BC101861.1 - 2 - 3 380 63

View Full Product Details