Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppp2r3a
Entrez Gene ID
363122
Gene Name
protein phosphatase 2, regulatory subunit B'', alpha
Gene Aliases
-
Chromosome Location
Chr.8: 109481352 - 109621460 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012202.2 NP_001012202.2 9 - 10 3230 112
XM_006243633.2 XP_006243695.1 10 - 11 3532 112
XM_006243634.2 XP_006243696.1 9 - 10 1956 112
GenBank mRNA BC087097.1 - 9 - 10 1305 112

View Full Product Details