Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Qrich1
Entrez Gene ID
301004
Gene Name
glutamine-rich 1
Gene Aliases
RGD1311095
Chromosome Location
Chr.8: 117305803 - 117346739 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134532.1 NP_001128004.1 7 - 8 2223 95
XM_006243744.2 XP_006243806.1 7 - 8 2223 95
XM_006243745.2 XP_006243807.1 7 - 8 3074 95
XM_006243746.2 XP_006243808.1 7 - 8 3086 95
XM_006243747.2 XP_006243809.1 6 - 7 3014 95
XM_006243749.2 XP_006243811.1 6 - 7 3026 95
XM_006243750.2 XP_006243812.1 5 - 6 1814 95
XM_008766506.1 XP_008764728.1 6 - 7 2646 95
XM_008766507.1 XP_008764729.1 6 - 7 2013 95

View Full Product Details