Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Qrich1
Entrez Gene ID
301004
Gene Name
glutamine-rich 1
Gene Aliases
RGD1311095
Chromosome Location
Chr.8: 117305803 - 117346739 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134532.1 NP_001128004.1 8 - 9 2328 68
XM_006243744.2 XP_006243806.1 8 - 9 2328 68
XM_006243745.2 XP_006243807.1 8 - 9 3179 68
XM_006243746.2 XP_006243808.1 8 - 9 3191 68
XM_006243747.2 XP_006243809.1 7 - 8 3119 68
XM_006243749.2 XP_006243811.1 7 - 8 3131 68
XM_006243750.2 XP_006243812.1 6 - 7 1919 68
XM_008766506.1 XP_008764728.1 7 - 8 2751 68
XM_008766507.1 XP_008764729.1 7 - 8 2118 68

View Full Product Details