Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Poc1a
Entrez Gene ID
501048
Gene Name
POC1 centriolar protein A
Gene Aliases
RGD1565004, Wdr51a
Chromosome Location
Chr.8: 114982341 - 115050844 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109296.1 NP_001102766.1 2 - 3 255 63
XM_008766561.1 XP_008764783.1 3 - 4 715 63
GenBank mRNA FQ233502.1 - 3 - 4 318 63

View Full Product Details