Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Poc1a
Entrez Gene ID
501048
Gene Name
POC1 centriolar protein A
Gene Aliases
RGD1565004, Wdr51a
Chromosome Location
Chr.8: 114982341 - 115050844 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109296.1 NP_001102766.1 4 - 5 544 94
XM_008766561.1 XP_008764783.1 5 - 6 1004 94
XM_008766562.1 XP_008764784.1 3 - 4 421 94
GenBank mRNA FQ233502.1 - 5 - 6 607 95

View Full Product Details