Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zcwpw2
Entrez Gene ID
681381
Gene Name
zinc finger, CW type with PWWP domain 2
Gene Aliases
-
Chromosome Location
Chr.8: 126268760 - 126402046 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766697.1 XP_008764919.1 3 - 4 1327 99

View Full Product Details