Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ripply2
Entrez Gene ID
363111
Gene Name
ripply transcriptional repressor 2
Gene Aliases
RGD1308746
Chromosome Location
Chr.8: 94676575 - 94691158 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006243504.2 XP_006243566.1 1 - 2 1371 84
XM_006243505.2 XP_006243567.1 2 - 3 251 84
XM_008766459.1 XP_008764681.1 1 - 2 1371 84
XM_343444.6 XP_343445.2 2 - 3 1134 84

View Full Product Details