Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc25a36
Entrez Gene ID
501039
Gene Name
solute carrier family 25 (pyrimidine nucleotide carrier ), member 36
Gene Aliases
-
Chromosome Location
Chr.8: 104962569 - 104995932 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766577.1 XP_008764799.1 1 - 2 252 59
XM_576451.6 XP_576451.4 1 - 2 251 59

Gene Symbol
Slc25a36l1
Entrez Gene ID
364991
Gene Name
solute carrier family 25 (pyrimidine nucleotide carrier ), member 36-like 1
Gene Aliases
RGD1561851
Chromosome Location
Chr.19: 37020220 - 37021997 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751866.3 XP_003751914.2 1 - 1 189 59

View Full Product Details