Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gnat1
Entrez Gene ID
363143
Gene Name
guanine nucleotide binding protein (G protein), alpha transducing activity polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.8: 116433302 - 116438038 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108780.2 NP_001102250.2 1 - 2 169 82

View Full Product Details