The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015017.1 | NP_001015017.1 | 5 - 6 | 911 | 75 |
XM_006242641.2 | XP_006242703.1 | 6 - 7 | 1203 | 75 | |
XM_006242642.2 | XP_006242704.1 | 5 - 6 | 2480 | 75 | |
XM_006242643.1 | XP_006242705.1 | 5 - 6 | 723 | 75 | |
GenBank mRNA | BC092647.1 | - | 5 - 6 | 911 | 75 |