The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057152.2 | NP_476493.2 | 5 - 6 | 2016 | 67 |
XM_006242591.2 | XP_006242653.1 | 5 - 6 | 2062 | 67 | |
XM_006242592.2 | XP_006242654.1 | 4 - 5 | 2563 | 67 | |
GenBank mRNA | AF304364.1 | - | 4 - 5 | 1782 | 67 |
FQ212383.1 | - | 5 - 6 | 2016 | 67 |