Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hnrnpk
Entrez Gene ID
117282
Gene Name
heterogeneous nuclear ribonucleoprotein K
Gene Aliases
Csbp, Hnrpk
Chromosome Location
Chr.17: 6664730 - 6676753 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057141.1 NP_476482.1 4 - 5 286 84
XM_006253552.2 XP_006253614.1 6 - 7 693 84
XM_006253553.2 XP_006253615.1 5 - 6 389 84
XM_006253554.2 XP_006253616.1 6 - 7 693 84
XM_006253555.2 XP_006253617.1 6 - 7 693 84
GenBank mRNA BC061867.1 - 5 - 6 373 84
D17711.1 - 4 - 5 286 84

View Full Product Details