The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053902.2 | NP_446354.1 | 13 - 14 | 1322 | 126 |
XM_006234138.2 | XP_006234200.1 | 12 - 13 | 1238 | 126 | |
GenBank mRNA | BC078762.1 | - | 13 - 14 | 1305 | 126 |
U68168.1 | - | 13 - 14 | 1323 | 126 |